Amido nero 10B ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulphonate, Amido Black 10B
A0586-25G
A0586-500G
A0586-100G
TCIAA0586-25GEA
25
EUR
TCIAA0586-25G
TCIAA0586-500G
TCIAA0586-100G
Amido nero 10B ≥95.0% (by HPLC)
Naphthalene black 10B (naphthol blue black, acid black 1)
Formula:
C₂₂H₁₄N₆Na₂O₉S₂ Peso molecolare: 616,49 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004017 CAS Number: 1064-48-8 EINECS: 213-903-1 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Lambda max. | 616.0 to 620.0 nm (H2O) |
Absorbance (E1%1cm) | min. 700 (H2O, 616.0 to 620.0 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...