Stampa…

N-(3-Aminopropyl)cyclohexylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

A0891-25ML A0891-500ML
TCIAA0891-25MLEA 40 EUR
TCIAA0891-25ML TCIAA0891-500ML
N-(3-Aminopropyl)cyclohexylamine ≥98.0% (by GC, titration analysis)
N-(3-Aminopropyl)cyclohexylamine
Formula: C₉H₂₀N₂
Peso molecolare: 156,27 g/mol
Punto di ebollizione: 123°C
Flash Pt: 101°C
MDL Number: MFCD00003838
CAS Number: 3312-60-5
EINECS: 222-001-7
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b350171fd147ae71c9fd9f1c1ef09fc4f196c804&vid=b350171fd147ae71c9fd9f1c1ef09fc4f196c804&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR