Stampa…

2-Mercaptoethanesulphonic acid sodium salt (mesna) ≥95.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

M0913-10G M0913-25G
TCIAM0913-10GEA 76 EUR
TCIAM0913-10G TCIAM0913-25G
2-Mercaptoethanesulphonic acid sodium salt (mesna) ≥95.0% (by titrimetric analysis)
2-Mercaptoethanesulphonic acid sodium salt (mesna)
Formula: C₂H₅NaO₃S₂
MDL Number: MFCD00007535
CAS Number: 19767-45-4
EINECS: 243-285-9
Merck Index: 14,05909
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6061cb4bc990f10bf9bdbfd782a8ea5824a6a97&vid=d6061cb4bc990f10bf9bdbfd782a8ea5824a6a97&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR