Stampa…

(±)-2-Methylglutaric acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Methylpentanedioic acid

M1300-25G M1300-5G
TCIAM1300-25GEA 192 EUR
TCIAM1300-25G TCIAM1300-5G
(±)-2-Methylglutaric acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylglutaric acid
Formula: C₆H₁₀O₄
Peso molecolare: 146,14 g/mol
Punto di ebollizione: 205°C
MDL Number: MFCD00002661
CAS Number: 617-62-9
EINECS: 210-521-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7155e9d6b318bbe1c1fa833c9e1bbb5d9c7f90f4&vid=7155e9d6b318bbe1c1fa833c9e1bbb5d9c7f90f4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR