Stampa…

2,2'-Bipyridine-4,4'-dicarboxylic acid ≥96.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

B1876-1G B1876-100MG
TCIAB1876-1GEA 59 EUR
TCIAB1876-1G TCIAB1876-100MG
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥96.0% (by HPLC, titration analysis)
2,2'-Bipyridine-4,4'-dicarboxylic acid
Formula: C₁₂H₈N₂O₄
Peso molecolare: 244,20 g/mol
MDL Number: MFCD00015430
CAS Number: 6813-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=265f1fbb64d6c87c24f230c6dbea924f33a6296c&vid=265f1fbb64d6c87c24f230c6dbea924f33a6296c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR