Diphenylphosphinic chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Diphenylphosphinyl chloride
Formula:
C₁₂H₁₀ClOP Peso molecolare: 236.63 g/mol Punto di ebollizione: 222°C Flash Pt: 26°C |
MDL Number:
MFCD00002077 CAS Number: 1499-21-4 EINECS: 216-107-2 UN: 2920 ADR: 8 II |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...