Stampa…

Tricyclohexylphosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

T2585-1G T2585-5G
TCIAT2585-1GEA 20 EUR
TCIAT2585-1G TCIAT2585-5G
Tricyclohexylphosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Tricyclohexylphosphonium tetrafluoroborate
Formula: C₁₈H₃₄BF₄P
Peso molecolare: 368,24 g/mol
MDL Number: MFCD03840580
CAS Number: 58656-04-5
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ab49709982e6b2c5721df45e863a5ce165b378f&vid=2ab49709982e6b2c5721df45e863a5ce165b378f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR