Stampa…

(+)-Dibenzoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid

D3826-25G D3826-250G
TCIAD3826-25GEA 22 EUR
TCIAD3826-25G TCIAD3826-250G
(+)-Dibenzoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
(+)-Dibenzoyl-D-tartaric acid
Formula: C₁₈H₁₄O₈
Peso molecolare: 358,31 g/mol
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c4cf0481c294d5f77230a856ece4ac28e42c2b&vid=82c4cf0481c294d5f77230a856ece4ac28e42c2b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR