Stampa…

(-)-Dibenzoyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

D3492-25G D3492-250G
TCIAD3492-25GEA 17 EUR
TCIAD3492-25G TCIAD3492-250G
(-)-Dibenzoyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
(-)-Dibenzoyl-L-tartaric acid
Formula: C₁₈H₁₄O₈
Peso molecolare: 358,31 g/mol
Punto di ebollizione: 234°C
MDL Number: MFCD00003074
CAS Number: 2743-38-6
EINECS: 220-374-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96ddbb10c8b12bcef6e2a0ad945dbf3f8189309d&vid=96ddbb10c8b12bcef6e2a0ad945dbf3f8189309d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR