Stampa…

o-Cresolphthalein

Fornitore: TCI EUROPE

Synonyms: 3,3-Bis(4-hydroxy-3-methylphenyl)isobenzofuran-1(3H)-one

C0404-25G C0404-100G
TCIAC0404-25GEA 40 EUR
TCIAC0404-25G TCIAC0404-100G
o-Cresolphthalein
o-Cresolphthalein
Formula: C₂₂H₁₈O₄
Peso molecolare: 346,39 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005912
CAS Number: 596-27-0
EINECS: 209-881-8
Merck Index: 14,02576
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e537496f76c1d124b58a17eff3ab6eb71714ea7c&vid=e537496f76c1d124b58a17eff3ab6eb71714ea7c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Lambda max. 563.0 to 568.0 nm (pH 9.8 buffer sol.)
Absorbance min. 0.590 (5mg/L, pH 9.8 buffer sol., 563.0 to 568.0 nm)
Solubility in EtOH almost transparency
Drying loss max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR