Cyclopentylacetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Cyclopentylacetic acid
C0894-25G
C0894-5G
TCIAC0894-25GEA
345
EUR
TCIAC0894-25G
TCIAC0894-5G
Cyclopentylacetic acid ≥98.0% (by GC, titration analysis)
Cyclopentylacetic acid
Formula:
C₇H₁₂O₂ Peso molecolare: 128,17 g/mol Punto di ebollizione: 231°C Flash Pt: 109°C |
MDL Number:
MFCD00001387 CAS Number: 1123-00-8 EINECS: 214-368-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...