Dipentyl phthalate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Di-n-pentyl phthalate
P0291-25G
P0291-500G
P0291-100G
TCIAP0291-25GEA
34
EUR
TCIAP0291-25G
TCIAP0291-500G
TCIAP0291-100G
Dipentyl phthalate ≥98.0% (by GC)
Dipentyl phthalate
Formula:
C₁₈H₂₆O₄ Peso molecolare: 306,41 g/mol Punto di ebollizione: 342°C Flash Pt: 118°C |
MDL Number:
MFCD00041934 CAS Number: 131-18-0 EINECS: 205-017-9 UN: 3082 ADR: 9,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...