Stampa…

2-Ethylhexyl mercaptoacetate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Thioglycolic acid 2-Ethylhexyl ester, Thioglycolic acid Octyl ester, Octyl Thioglycolate, Mercaptoacetic acid 2-Ethylhexyl ester, 2-Ethylhexyl Mercaptoacetate, 2-Ethylhexyl Thioglycolate

T0612-25ML T0612-500ML
TCIAT0612-25MLEA 16 EUR
TCIAT0612-25ML TCIAT0612-500ML
2-Ethylhexyl mercaptoacetate ≥98.0% (by GC)
2-Ethylhexyl mercaptoacetate
Formula: C₁₀H₂₀O₂S
Peso molecolare: 204,3296 g/mol
Flash Pt: 136°C
MDL Number: MFCD00022081
CAS Number: 7659-86-1
EINECS: 231-626-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2953ffd3c98240810665a7e4dac1a87005b14b39&vid=2953ffd3c98240810665a7e4dac1a87005b14b39&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR