Stampa…

5-Fluoroorotic acid monohydrate (5-FOA) ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-FOA

F0382-1G F0382-100MG
TCIAF0382-1GEA 131 EUR
TCIAF0382-1G TCIAF0382-100MG
5-Fluoroorotic acid monohydrate (5-FOA) ≥95.0% (by HPLC, titration analysis)
5-Fluoroorotic acid monohydrate (5-FOA)
Formula: C₅H₃FN₂O₄·1H₂O
Peso molecolare: 174,0 g/mol (anhy)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00150658
CAS Number: 220141-70-8
EINECS: 211-876-0
Merck Index: 14,04176
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c769e34212d77320c9cafd1aecbd0a31225f7fe&vid=6c769e34212d77320c9cafd1aecbd0a31225f7fe&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 % (calcd.on anh.substance)
Solubility in 1mol/L NaOH faint turbidity
Water 8.5 to 10.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR