Calcium cyanamide
Fornitore: TCI
|
Pericolo
|
C0006-25G
C0006-500G
TCIAC0006-25GEA
16
EUR
TCIAC0006-25G
TCIAC0006-500G
Calcium cyanamide
Calcium cyanamide
Formula:
CCaN₂ Peso molecolare: 80.11 g/mol |
MDL Number:
MFCD00064894 CAS Number: 156-62-7 EINECS: 205-861-8 UN: 1403 ADR: 4.3 III Merck Index: 14,01662 |
|
Risultati del test di specificazione
Appearance | Light gray to Brown to Black powder to crystal |
Nitorogen of cyanamide | min. 19.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...