Stampa…

3,6-Dithia-1,8-octanediol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,8-Dihydroxy-3,6-Dithiaoctane, 2,2-(Ethane-1,2-diylbis(sulphanediyl))diethanol

D2188-25G D2188-250G
TCIAD2188-25GEA 22 EUR
TCIAD2188-25G TCIAD2188-250G
3,6-Dithia-1,8-octanediol ≥98.0% (by GC)
3,6-Dithia-1,8-octanediol
Formula: C₆H₁₄O₂S₂
Peso molecolare: 182,31 g/mol
Punto di ebollizione: 100°C
MDL Number: MFCD00002911
CAS Number: 5244-34-8
EINECS: 226-045-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f518e252879c302b5045c4f170db3253e5da457&vid=4f518e252879c302b5045c4f170db3253e5da457&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 67.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR