Stampa…

N,N-Diethylacetoacetamide ≥98.0% (by GC)

Fornitore: TCI

D1371-25G D1371-500G
TCIAD1371-25GEA 32 EUR
TCIAD1371-25G TCIAD1371-500G
N,N-Diethylacetoacetamide ≥98.0% (by GC)
N,N-Diethylacetoacetamide
Formula: C₈H₁₅NO₂
Punto di ebollizione: 116°C
Flash Pt: 120°C
MDL Number: MFCD00026728
CAS Number: 2235-46-3
EINECS: 218-792-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4001bbc98c9beb322bafa71efd3d406c7c90e99f&vid=4001bbc98c9beb322bafa71efd3d406c7c90e99f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR