Methyl (methoxymethylene)acetate ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Methyl 3-methoxyacrylate
M0995-25ML
M0995-500ML
TCIAM0995-25MLEA
27
EUR
TCIAM0995-25ML
TCIAM0995-500ML
Methyl (methoxymethylene)acetate ≥95.0% (by GC)
Methyl (methoxymethylene)acetate
Formula:
C₅H₈O₃ Peso molecolare: 116,1152 g/mol Punto di ebollizione: 56°C Flash Pt: 65°C |
MDL Number:
MFCD00128206 CAS Number: 34846-90-7 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...