Stampa…

1-(4-Bromophenyl)pyrrolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-(4-Bromophenyl)pyrrolidine

B3900-1G B3900-5G
TCIAB3900-1GEA 23 EUR
TCIAB3900-1G TCIAB3900-5G
1-(4-Bromophenyl)pyrrolidine ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)pyrrolidine
Formula: C₁₀H₁₂BrN
Peso molecolare: 226,1130 g/mol
MDL Number: MFCD04112480
CAS Number: 22090-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd324f46005ebea0df48605bd54c4ef77eb6ff5&vid=1cd324f46005ebea0df48605bd54c4ef77eb6ff5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR