Stampa…

3-Iodo-L-tyrosine ≥98.0% (by titrimetric analysis)

Fornitore: TCI

I0075-1G I0075-5G
TCIAI0075-1GEA 55 EUR
TCIAI0075-1G TCIAI0075-5G
3-Iodo-L-tyrosine ≥98.0% (by titrimetric analysis)
3-Iodo-L-tyrosine
Formula: C₉H₁₀INO₃
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002608
CAS Number: 70-78-0
EINECS: 200-744-8
Merck Index: 14,05047
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a26782b60b8c57594d7f41bb26d232e0e7d29057&vid=a26782b60b8c57594d7f41bb26d232e0e7d29057&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -2.0 to -5.0 deg (C=2, 1mol/L HCl)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR