2-Chloro-3-methoxybenzaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
C2185-25G
C2185-5G
TCIAC2185-25GEA
137
EUR
TCIAC2185-25G
TCIAC2185-5G
2-Chloro-3-methoxybenzaldehyde ≥98.0% (by GC)
2-Chloro-3-methoxybenzaldehyde
Formula:
C₈H₇ClO₂ Peso molecolare: 170,5930 g/mol Punto di fusione: 53°C |
MDL Number:
MFCD07369787 CAS Number: 54881-49-1 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...