Stampa…

3-Hydroxy-3-methyl butyric acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid

H0701-25G H0701-5G
TCIAH0701-25GEA 188 EUR
TCIAH0701-25G TCIAH0701-5G
3-Hydroxy-3-methyl butyric acid ≥96.0% (by GC, titration analysis)
3-Hydroxy-3-methyl butyric acid
Formula: C₅H₁₀O₃
Peso molecolare: 118,13 g/mol
MDL Number: MFCD00059081
CAS Number: 625-08-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR