4-Bromo-p-terphenyl ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B2155-1G
B2155-5G
TCIAB2155-1GEA
123
EUR
TCIAB2155-1G
TCIAB2155-5G
4-Bromo-p-terphenyl ≥97.0% (by GC)
4-Bromo-p-terphenyl
Formula:
C₁₈H₁₃Br Peso molecolare: 309,1998 g/mol Punto di fusione: 232°C |
MDL Number:
MFCD02685117 CAS Number: 1762-84-1 UN: 3152 ADR: 9,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...