(S)-(+)-4-Phenyl-2-oxazolidinone ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-4-Phenyloxazolidin-2-one
P1308-25G
P1308-5G
TCIAP1308-25GEA
151
EUR
TCIAP1308-25G
TCIAP1308-5G
(S)-(+)-4-Phenyl-2-oxazolidinone ≥99.0% (by GC)
(S)-(+)-4-Phenyl-2-oxazolidinone
Formula:
C₉H₉NO₂ Peso molecolare: 163,18 g/mol |
MDL Number:
MFCD00043396 CAS Number: 99395-88-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 130.0 to 133.0 deg-C |
Specific rotation [a]20/D | +70.0 to +74.0 deg (C=1, EtOAc) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...