Stampa…

(S)-(-)-4-Isopropyl-2-oxazolidinone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one

I0451-1G I0451-25G I0451-5G
TCIAI0451-1GEA 16 EUR
TCIAI0451-1G TCIAI0451-25G TCIAI0451-5G
(S)-(-)-4-Isopropyl-2-oxazolidinone ≥98.0% (by GC)
(S)-(-)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
Peso molecolare: 129,16 g/mol
MDL Number: MFCD00010847
CAS Number: 17016-83-0
EINECS: 469-920-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee85b8af0d9c637c7db47422fbd2ef7467c0903f&vid=ee85b8af0d9c637c7db47422fbd2ef7467c0903f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 73.0 deg-C
Specific rotation [a]20/D -15.0 to -20.0 deg (C=6, EtOH)
Solubility in EtOH very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR