Stampa…

(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

I0572-1G I0572-25G I0572-5G
TCIAI0572-1GEA 24 EUR
TCIAI0572-1G TCIAI0572-25G TCIAI0572-5G
(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98.0% (by GC)
(R)-(+)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
Peso molecolare: 129,16 g/mol
MDL Number: MFCD00075172
CAS Number: 95530-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f007158399659d17f22d0a99f452460cc00822&vid=65f007158399659d17f22d0a99f452460cc00822&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 74.0 deg-C
Specific rotation [a]20/D +15.0 to +20.0 deg (C=6, EtOH)
Solubility in EtOH very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR