Stampa…

N-Phenyl-2-naphthylamine ≥97.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

P0198-25G P0198-500G
TCIAP0198-25GEA 18 EUR
TCIAP0198-25G TCIAP0198-500G
N-Phenyl-2-naphthylamine ≥97.0% (by total nitrogen basis)
N-Phenyl-2-naphthylamine
Formula: C₁₆H₁₃N
Peso molecolare: 219,29 g/mol
Punto di ebollizione: 237°C
MDL Number: MFCD00004052
CAS Number: 135-88-6
EINECS: 205-223-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a476efd6bca636216eaa41be1abb3658ca332b29&vid=a476efd6bca636216eaa41be1abb3658ca332b29&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 108.0 to 110.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR