Stampa…

N-Phenyl-1-naphthylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Anilinonaphthalene

P0197-25G P0197-500G
TCIAP0197-25GEA 19 EUR
TCIAP0197-25G TCIAP0197-500G
N-Phenyl-1-naphthylamine ≥98.0% (by GC)
N-Phenyl-1-naphthylamine
Formula: C₁₆H₁₃N
Peso molecolare: 219,29 g/mol
Punto di ebollizione: 200°C
MDL Number: MFCD00003878
CAS Number: 90-30-2
EINECS: 201-983-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf8e6228b67337c5696bad38db58322e3f2c5b06&vid=bf8e6228b67337c5696bad38db58322e3f2c5b06&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR