Bis(4-hydroxyphenyl)disulphide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4'-Dithiodiphenol, 4,4'-Dihydroxydiphenyl disulphide, Bis(4-hydroxyphenyl) Disulphide
B3827-5G
B3827-1G
TCIAB3827-5GEA
87
EUR
TCIAB3827-5G
TCIAB3827-1G
Bis(4-hydroxyphenyl)disulphide ≥98.0% (by GC)
Bis(4-hydroxyphenyl)disulphide
Formula:
C₁₂H₁₀O₂S₂ Peso molecolare: 250,34 g/mol |
MDL Number:
MFCD00020164 CAS Number: 15015-57-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...