Hexachlorophene ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,2'-Methylenebis(3,4,6-trichlorophenol), 3,3',5,5',6,6'-Hexachloro-2,2'-dihydroxydiphenylmethane
M0219-1G
M0219-5G
TCIAM0219-1GEA
41
EUR
TCIAM0219-1G
TCIAM0219-5G
Hexachlorophene ≥98.0% (by titrimetric analysis)
Hexachlorophene
Formula:
C₁₃H₆Cl₆O₂ Peso molecolare: 406,91 g/mol |
MDL Number:
MFCD00002171 CAS Number: 70-30-4 EINECS: 200-733-8 UN: 2875 ADR: 6.1,III Merck Index: 14,04680 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 162.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...