Stampa…

1-Methyl-1-phenylhydrazine ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M0399-25G M0399-5G
TCIAM0399-25GEA 193 EUR
TCIAM0399-25G TCIAM0399-5G
1-Methyl-1-phenylhydrazine ≥95.0% (by GC, titration analysis)
1-Methyl-1-phenylhydrazine
Formula: C₇H₁₀N₂
Punto di ebollizione: 109°C
Flash Pt: 92°C
MDL Number: MFCD00007627
CAS Number: 618-40-6
EINECS: 210-547-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31c7a61079646b8513b17039d17a464c77c9ad54&vid=31c7a61079646b8513b17039d17a464c77c9ad54&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR