Stampa…

Disodium terephthalate ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: Terephthalic acid disodium salt

T1097-500G T1097-25G T1097-100G
TCIAT1097-500GEA 360 EUR
TCIAT1097-500G TCIAT1097-25G TCIAT1097-100G
Disodium terephthalate ≥99.0% (by HPLC, titration analysis)
Disodium terephthalate
Formula: C₈H₄Na₂O₄
MDL Number: MFCD00013137
CAS Number: 10028-70-3
EINECS: 233-075-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f11c17d2966824b8483de05ec85c2efbd9f0d423&vid=f11c17d2966824b8483de05ec85c2efbd9f0d423&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR