trans-Aconitic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid
A0127-25G
A0127-500G
TCIAA0127-25GEA
46
EUR
TCIAA0127-25G
TCIAA0127-500G
trans-Aconitic acid ≥98.0% (by HPLC, titration analysis)
trans-Aconitic acid
Formula:
C₆H₆O₆ Peso molecolare: 174,11 g/mol |
MDL Number:
MFCD00002721 CAS Number: 4023-65-8 EINECS: 223-688-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...