Fumaric acid monoethyl ester ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
(E)-4-Ethoxy-4-oxobut-2-enoic acid, Monoethyl fumarate, Ethyl hydrogen fumarate
Formula:
C₆H₈O₄ Peso molecolare: 144.13 g/mol |
MDL Number:
MFCD00002699 CAS Number: 2459-05-4 EINECS: 219-544-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 67.0 to 70.0 deg-C |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...