Sodium hippurate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Hippuric Acid Sodium Salt, Sodium 2-benzamidoacetate
Formula:
C₉H₈NNaO₃ Peso molecolare: 201.16 g/mol |
MDL Number:
MFCD00002693 CAS Number: 532-94-5 EINECS: 208-548-4 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...