3-Methyl-2-oxovaleric acid ≥95.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Methyl-2-oxopentanoic acid
K0020-1G
K0020-5G
TCIAK0020-1GEA
137
EUR
TCIAK0020-1G
TCIAK0020-5G
3-Methyl-2-oxovaleric acid ≥95.0% (by GC, titration analysis)
3-Methyl-2-oxovaleric acid
Formula:
C₆H₁₀O₃ Peso molecolare: 130,14 g/mol Punto di ebollizione: 84°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002582 CAS Number: 1460-34-0 EINECS: 215-955-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 38.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...