Stampa…

2-Chloro-N-(2,6-dimethylphenyl)acetamide ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide

C2307-25G C2307-5G
TCIAC2307-25GEA 69 EUR
TCIAC2307-25G TCIAC2307-5G
2-Chloro-N-(2,6-dimethylphenyl)acetamide ≥98.0% (by GC, total nitrogen)
2-Chloro-N-(2,6-dimethylphenyl)acetamide
Formula: C₁₀H₁₂ClNO
Peso molecolare: 197,66 g/mol
MDL Number: MFCD00000926
CAS Number: 1131-01-7
EINECS: 214-460-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dacbf72634bbb7612043e77d34090411461b8c0&vid=2dacbf72634bbb7612043e77d34090411461b8c0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR