Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Nitrilotriacetic acid, trisodium salt, monohydrate
N0101-25G
N0101-500G
TCIAN0101-25GEA
16
EUR
TCIAN0101-25G
TCIAN0101-500G
Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% (by titrimetric analysis)
Nitrilotriacetic acid trisodium salt monohydrate
Formula:
C₆H₆NNa₃O₆·1H₂O Peso molecolare: 275,1 g/mol |
MDL Number:
MFCD00150789 CAS Number: 18662-53-8 EINECS: 225-768-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
pH | 10.0 to 12.0 (50 g/L, 25 deg-C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...