Stampa…

N-Acetyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

A0693-25G A0693-250G
TCIAA0693-25GEA 20 EUR
TCIAA0693-25G TCIAA0693-250G
N-Acetyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)
N-Acetyl-L-glutamic acid
Formula: C₇H₁₁NO₅
Peso molecolare: 189,17 g/mol
MDL Number: MFCD00002802
CAS Number: 1188-37-0
EINECS: 214-708-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c1e0de64734960d13938d481e5da3b5ae4a7365&vid=1c1e0de64734960d13938d481e5da3b5ae4a7365&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -13.0 to -17.0 deg (C=1, H2O)
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR