DL-Lactamide ≥98.0% (by total nitrogen basis)
Fornitore: TCI
Synonyms:
2-Hydroxypropionamide, Lactamide
Formula:
C₃H₇NO₂ Peso molecolare: 89.09 g/mol |
MDL Number:
MFCD00008017 CAS Number: 2043-43-8 EINECS: 218-049-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 77.0 to 80.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...