Diethyl allylmalonate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₆O₄ Flash Pt: 92°C |
MDL Number:
MFCD00009155 CAS Number: 2049-80-1 EINECS: 218-072-9 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...