Stampa…

4-Quinazolone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one

H0803-25G H0803-5G
TCIAH0803-25GEA 53 EUR
TCIAH0803-25G TCIAH0803-5G
4-Quinazolone ≥98.0% (by titrimetric analysis)
4-Quinazolone
Formula: C₈H₆N₂O
Peso molecolare: 146,15 g/mol
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83ddbc15a1bd0667a9e3d3aa99f368c26ac34f27&vid=83ddbc15a1bd0667a9e3d3aa99f368c26ac34f27&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 215.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR