4-Nitro-2,1,3-benzothiadiazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Nitrobenzo[c][1,2,5]thiadiazole
N0786-25G
N0786-5G
TCIAN0786-25GEA
211
EUR
TCIAN0786-25G
TCIAN0786-5G
4-Nitro-2,1,3-benzothiadiazole ≥98.0% (by GC)
4-Nitro-2,1,3-benzothiadiazole
Formula:
C₆H₃N₃O₂S Peso molecolare: 181,17 g/mol |
MDL Number:
MFCD00068050 CAS Number: 6583-06-8 EINECS: 229-514-5 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...