3-Methylthiophene-2-carboxaldehyde ≥85.0% (by GC)
Fornitore: TCI
Formula:
C₆H₆OS Punto di ebollizione: 113°C Flash Pt: 82°C |
MDL Number:
MFCD00005430 CAS Number: 5834-16-2 EINECS: 227-418-8 |
|
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...