1,4-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2'-(1,4-Phenylene)diacetic acid
P1433-25G
P1433-5G
TCIAP1433-25GEA
235
EUR
TCIAP1433-25G
TCIAP1433-5G
1,4-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)
1,4-Phenylenediacetic acid
Formula:
C₁₀H₁₀O₄ Peso molecolare: 194,19 g/mol |
MDL Number:
MFCD00004354 CAS Number: 7325-46-4 EINECS: 230-801-2 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...