1,3-Phenylenediacetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,2'-(1,3-Phenylene)diacetic acid
Formula:
C₁₀H₁₀O₄ Peso molecolare: 194.19 g/mol |
MDL Number:
MFCD00004341 CAS Number: 19806-17-8 EINECS: 243-332-3 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...