Stampa…

Trimellitic anhydride chloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride

T1079-25G T1079-500G T1079-100G
TCIAT1079-25GEA 19 EUR
TCIAT1079-25G TCIAT1079-500G TCIAT1079-100G
Trimellitic anhydride chloride ≥98.0% (by titrimetric analysis)
Trimellitic anhydride chloride
Formula: C₉H₃ClO₄
Peso molecolare: 210,57 g/mol
MDL Number: MFCD00005924
CAS Number: 1204-28-0
EINECS: 214-874-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e690883a58fb3cf6edcec801f8dc818a61c18b2&vid=4e690883a58fb3cf6edcec801f8dc818a61c18b2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 67.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR