Stampa…

Bis(methylthio)methane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B1204-25ML B1204-5ML
TCIAB1204-25MLEA 71 EUR
TCIAB1204-25ML TCIAB1204-5ML
Bis(methylthio)methane ≥98.0% (by GC)
Bis(methylthio)methane
Formula: C₃H₈S₂
Peso molecolare: 108,23 g/mol
Punto di ebollizione: 152°C
Flash Pt: 44°C
MDL Number: MFCD00008564
CAS Number: 1618-26-4
EINECS: 216-577-9
UN: 1993
ADR: 3,III
Merck Index: 14,01256
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b2a70242fd02f030bd7fde880f1eef8f78f380&vid=04b2a70242fd02f030bd7fde880f1eef8f78f380&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR