Acido α-chetoglutarico sale disodico ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Disodium 2-Ketoglutate, Disodium 2-Oxoglutarate, 2-Ketoglutaric Acid Disodium Salt
Formula:
C₅H₄Na₂O₅ Peso molecolare: 190.06 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00043347 CAS Number: 305-72-6 EINECS: 206-167-8 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...