Stampa…

2-Amino-4-hydroxy-6-methylpyrimidine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol

A1204-25G A1204-250G
TCIAA1204-25GEA 29 EUR
TCIAA1204-25G TCIAA1204-250G
2-Amino-4-hydroxy-6-methylpyrimidine ≥98.0% (by HPLC, titration analysis)
2-Amino-4-hydroxy-6-methylpyrimidine
Formula: C₅H₇N₃O
Peso molecolare: 125,13 g/mol
MDL Number: MFCD00006095
CAS Number: 3977-29-5
EINECS: 223-612-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1fb69f307c47cb335372982c48222316a8f80ed&vid=c1fb69f307c47cb335372982c48222316a8f80ed&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in HCl (1+3) almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR